Learn More
Medchemexpress LLC Nepsilon-acetyl-L-lysine-d8 | 98.5% | 196.27 | 1 MG
SDP
Catalog No. 5000435563
Encompass_Preferred
Nepsilon-acetyl-L-lysine-d8 is the deuterium labeled Nepsilon-Acetyl-L-lysine. Nepsilon-Acetyl-L-lysine is an endogenous metabolite and an R-chain N-acetylated α amino acid. Stable heavy isotopes are incorporated into drug molecules primarily as tracers for quantitation during drug development, and deuteration has garnered attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Functions as an endogenous metabolite.
  • Functions as an R-chain N-acetylated α amino acid.
  • May affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-35563 Supplier Medchemexpress LLC Supplier No. HY113426S1MG
Nepsilon-acetyl-L-lysine-d8 is the deuterium labeled Nepsilon-Acetyl-L-lysine. Nepsilon-Acetyl-L-lysine is an endogenous metabolite and an R-chain N-acetylated α amino acid. Stable heavy isotopes are incorporated into drug molecules primarily as tracers for quantitation during drug development, and deuteration has garnered attention due to its potential impact on the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Functions as an endogenous metabolite.
  • Functions as an R-chain N-acetylated α amino acid.
  • May affect pharmacokinetic and metabolic profiles of drugs.