Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-L-lysine, 95%

Catalog No. AAH6376803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6376803 1 g
AAH6376806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6376803 Supplier Thermo Scientific Chemicals Supplier No. H6376803
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 146982-27-6 | C25H28N2O6 | 452.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146982-27-6
Molecular Formula C25H28N2O6
Molecular Weight (g/mol) 452.51
MDL Number MFCD00190872
InChI Key OJBNDXHENJDCBA-ANBDAQEENA-N
Synonym fmoc-lys alloc-oh, fmoc-lys aloc-oh, n-fmoc-n'-allyoxycarbonyl-l-lysine, s-2-9h-ffuoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-allyloxy carbonyl amino hexanoic acid, fmoc-lys alloc, aloc-lys fmoc-oh, pubchem13153, fmoc-l-lys alloc-oh, n, a-fmoc-n, a-alloc-l-lysine
PubChem CID 11123257
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(prop-2-enoxycarbonylamino)hexanoic acid
SMILES OC(=O)[C@H](CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Beilstein 5893081
Quantity 1 g
Formula Weight 452.5
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Nepsilon-Allyloxycarbonyl-Nalpha-Fmoc-L-lysine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.