Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride, 98%, Thermo Scientific™

Catalog No. AAH6371406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6371406 5 g
AAH6371403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6371406 Supplier Thermo Scientific Chemicals Supplier No. H6371406

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6366-70-7
Molecular Formula C21H27ClN2O4
Molecular Weight (g/mol) 406.91
MDL Number MFCD00038981
InChI Key NWSFLBWTZZJOAF-FYZYNONXSA-N
Synonym h-lys z-obzl.hcl, n6-cbz-l-lysine benzyl ester hydrochloride, h-lys z-obzl hydrochloride, s-benzyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, h-lys z-obzl hcl, h-lys z-obzl inverted exclamation mark currencyhcl, lys z-obzl.hcl, benzyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, s-benzyl 2-amino-6-benzyloxy carbonyl amino hexanoate hcl, lys z-obzl hcl
PubChem CID 11750132
IUPAC Name benzyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride
SMILES Cl.N[C@@H](CCCCNOC(=O)CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Color White
Beilstein 3580519
Quantity 5 g
Formula Weight 406.9
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-L-lysine benzyl ester hydrochloride
EINECSNumber 228-859-9
RTECSNumber OL5685000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.