Learn More
Andwin Scientific NEPTNE FLTER TPS 100UL BT100.X
SDP
Catalog No. 501126025
Encompass_Preferred
The 100 L Barrier Pipet Tips by Neptune are specially designed with high density polyethlene filters to avoid sample contamination and with S3 polymer tips to help remove sample hold up. It is sold in a pack of 960 tips.
Catalog No. 50-112-6025 Supplier Andwin Scientific Supplier No. 89140162
The 100 L Barrier Pipet Tips by Neptune are specially designed with high density polyethlene filters to avoid sample contamination and with S3 polymer tips to help remove sample hold up. It is sold in a pack of 960 tips.