Learn More
Andwin Scientific NEPTUNE BT BARRIER PIPET TIPS
SDP
Catalog No. NC0327518
Encompass_Preferred
The Neptune BT Barrier Pipet Tips are constructed from virgin polypropylene. This product hinders aerosol contamination and offer excellent consistency. It features a thin wall design that gives an optimal seal for both single and multichannel pipettors. The tips have a universal fit and low retention. These have a volume capacity of 200 L and S3 Polymer Tips that remove sample hold up and guarantee a more precise drawing and dispensing of samples. This product is sold in a case of 4800 tips.
Catalog No. NC0327518 Supplier Andwin Scientific Supplier No. 89140936CS
The Neptune BT Barrier Pipet Tips are constructed from virgin polypropylene. This product hinders aerosol contamination and offer excellent consistency. It features a thin wall design that gives an optimal seal for both single and multichannel pipettors. The tips have a universal fit and low retention. These have a volume capacity of 200 L and S3 Polymer Tips that remove sample hold up and guarantee a more precise drawing and dispensing of samples. This product is sold in a case of 4800 tips.