Learn More
MSC Kwazar Backpack Sprayer, Electric, 15 L, High Density Polyethylene
Catalog No. 502837301
Encompass
This electric backpack sprayer is designed for demanding users, offering consistent pressure for liquid application. Its battery allows for spraying approximately 150 liters of liquid over 8-10 hours. Ideal for garden plants, crops, fruit trees, and pest control.
  • Efficient battery: 8-10 hours run time.
  • Comfortable carrying system.
  • Adjustable fiberglass lance (0.6-1.2 m spray range).
  • Pressure gauge on handle.
  • Tank with 6-year warranty.
  • Easy compression action.
  • Durable, chemical-resistant components.
  • Hidden pump unit ensures safety and constant pressure.
  • Includes strap, nozzle, battery, wand, charger.
  • In-tank filter.
Catalog No. 50-283-7301 Supplier MSC Supplier No. 74335654
This electric backpack sprayer is designed for demanding users, offering consistent pressure for liquid application. Its battery allows for spraying approximately 150 liters of liquid over 8-10 hours. Ideal for garden plants, crops, fruit trees, and pest control.
  • Efficient battery: 8-10 hours run time.
  • Comfortable carrying system.
  • Adjustable fiberglass lance (0.6-1.2 m spray range).
  • Pressure gauge on handle.
  • Tank with 6-year warranty.
  • Easy compression action.
  • Durable, chemical-resistant components.
  • Hidden pump unit ensures safety and constant pressure.
  • Includes strap, nozzle, battery, wand, charger.
  • In-tank filter.