Learn More
MSC Kwazar Backpack Sprayer, 15 Liter, FPM Seal, Chemical Resistant
Catalog No. 502836001
Encompass
The Neptune Pro+ Backpack Sprayer is designed for efficient and precise application in large-scale gardening and agricultural settings. This 15 liter sprayer features an ergonomic design with a unique no-leak tank, ensuring comfortable and safe operation. Its components are constructed from highly durable and chemical-resistant materials, making it suitable for a wide range of substances.
  • Easy-to-use compression action.
  • FPM seal resistant to most common chemicals and detergents.
  • Ergonomic design with a unique, no-leak tank.
  • Hidden pump unit ensures safety and constant pressure in the lance for precise spraying.
  • Includes a telescopic lance, pressure gauge, and waist belt.
  • Ideal for large-scale gardening and agricultural cultivations.
  • Components made from highly durable and chemical-resistant materials.
Catalog No. 50-283-6001 Supplier MSC Supplier No. 74333451
The Neptune Pro+ Backpack Sprayer is designed for efficient and precise application in large-scale gardening and agricultural settings. This 15 liter sprayer features an ergonomic design with a unique no-leak tank, ensuring comfortable and safe operation. Its components are constructed from highly durable and chemical-resistant materials, making it suitable for a wide range of substances.
  • Easy-to-use compression action.
  • FPM seal resistant to most common chemicals and detergents.
  • Ergonomic design with a unique, no-leak tank.
  • Hidden pump unit ensures safety and constant pressure in the lance for precise spraying.
  • Includes a telescopic lance, pressure gauge, and waist belt.
  • Ideal for large-scale gardening and agricultural cultivations.
  • Components made from highly durable and chemical-resistant materials.