Learn More
TARGETMOL CHEMICALS INC NERATINIB 100MG
SDP
Catalog No. 502577159
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Neratinib (HKI-272) (HKI-272) is an orally available irreversible tyrosine kinase inhibitor for HER2 and EGFR (IC50 59/92 nM) respectively. purity: 98%
Catalog No. 50-257-7159 Supplier TARGETMOL CHEMICALS INC Supplier No. T2325100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg and bulk. Please contact Fisher for quotes. Neratinib (HKI-272) (HKI-272) is an orally available irreversible tyrosine kinase inhibitor for HER2 and EGFR (IC50 59/92 nM) respectively. purity: 98%