Learn More
Medchemexpress LLC HY-32721 10mg Medchemexpress, Neratinib CAS:698387-09-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY3272110MG
Encompass_Preferred
Medchemexpress, HY-32721 10mg Neratinib CAS:698387-09-6 Neratinib is an orally available, irreversible tyrosine kinase inhibitor with IC50s of 59 nM and 92 nM for HER2 and EGFR, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9310