Learn More
Medchemexpress LLC HY-32721 200mg , Neratinib CAS:698387-09-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY32721200MG
Encompass_Preferred
HY-32721 200mg Neratinib CAS: 698387-09-6 Neratinib (HKI-272) is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9791