Learn More
Medchemexpress LLC Neratinib-d6 | 1259519-18-0 | 563.08 | 1 MG
SDP
Catalog No. 5000404271
Encompass_Preferred
Neratinib-d6 (HKI-272-d6) is the deuterium labeled Neratinib. Neratinib (HKI-272) is an orally available, irreversible tyrosine kinase inhibitor with IC50s of 59 nM and 92 nM for HER2 and EGFR, respectively. For research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-04271 Supplier Medchemexpress LLC Supplier No. HY32721S1MG
Neratinib-d6 (HKI-272-d6) is the deuterium labeled Neratinib. Neratinib (HKI-272) is an orally available, irreversible tyrosine kinase inhibitor with IC50s of 59 nM and 92 nM for HER2 and EGFR, respectively. For research use only.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.