Learn More
Enzo Life Sciences Neratinib (free base) (100mg). CAS: 698387-09-6
SDP
Supplier:  Enzo Life Sciences ENZCHM2280100
Encompass_Preferred
Neratinib is a potent and selective inhibitor of EGFR and HER2. Alternative name: HKI-272. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO at 2 mg/mL. Long Term Storage: -20°C.
Catalog No. 50-201-3618