Learn More
Apexbio Technology LLC Neratinib (HKI-272), 25mg. Cas: 698387-09-6 MFCD: MFCD09752958. HER2/EGFR inhibitor,potent and irreversible
SDP
Supplier:  Apexbio Technology LLC A832225
Encompass_Preferred
IC50: 59 nM (Her-2); 92 nM (EGFR)HER-2 belongs to the ErbB family of receptor tyrosine kinases, which has been implicated in a variety of cancers. Overexpression of HER-2 is seen in 2530% of breast cancer patients and predicts a poor outcome in patients with primary disease. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5108