Learn More
Medchemexpress LLC Neratinib (maleate) | 915942-22-2 | 99.7% | 673.11 | 1 ML
SDP
Catalog No. 5000401584
Encompass_Preferred
Neratinib (HKI-272) maleate is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively. This product is for research use only.
  • Inhibits HER2 and EGFR with IC50s of 59 nM and 92 nM.
  • Shows anticancer activities against cancer cells expressing high levels of HER-2 or EGFR.
  • Reduces tumor growth in a dose-dependent manner in various xenografts.
  • Inhibits proliferation of cell lines with high HER-2 levels.
  • Arrests cell cycle at G1-S phase in BT474 cells.
  • Inhibits MAPK and Akt phosphorylation.
  • Down-regulates cyclin D1 levels and induces p27.
  • Inactive in MX-1 and MCF-7 xenografts.
  • Inhibited phosphorylation of HER-2 in BT474 xenografts.
Catalog No. 50-004-01584 Supplier Medchemexpress LLC Supplier No. HY32721B10MM/1ML
Neratinib (HKI-272) maleate is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively. This product is for research use only.
  • Inhibits HER2 and EGFR with IC50s of 59 nM and 92 nM.
  • Shows anticancer activities against cancer cells expressing high levels of HER-2 or EGFR.
  • Reduces tumor growth in a dose-dependent manner in various xenografts.
  • Inhibits proliferation of cell lines with high HER-2 levels.
  • Arrests cell cycle at G1-S phase in BT474 cells.
  • Inhibits MAPK and Akt phosphorylation.
  • Down-regulates cyclin D1 levels and induces p27.
  • Inactive in MX-1 and MCF-7 xenografts.
  • Inhibited phosphorylation of HER-2 in BT474 xenografts.