Learn More
Medchemexpress LLC Neratinib maleate | 915942-22-2 | 99.7% | 673.11 | 50 MG
SDP
Catalog No. 5000401585
Encompass_Preferred
Neratinib maleate (HKI-272 maleate) is an orally available, irreversible, highly selective HER2 and EGFR inhibitor. It is intended for research use only.
  • Inhibits HER2 and EGFR with IC50s of 59 nM and 92 nM, respectively.
  • Demonstrates antiproliferative effects in cell lines with high HER-2 levels.
  • Arrests cell cycle at G1-S phase in BT474 cells.
  • Inhibits MAPK and Akt phosphorylation, and induces p27.
  • Exhibits anticancer activities against HER-2 or EGFR expressing cancer cells in vivo.
  • Reduces tumor growth in xenograft models.
Catalog No. 50-004-01585 Supplier Medchemexpress LLC Supplier No. HY32721B50MG
Neratinib maleate (HKI-272 maleate) is an orally available, irreversible, highly selective HER2 and EGFR inhibitor. It is intended for research use only.
  • Inhibits HER2 and EGFR with IC50s of 59 nM and 92 nM, respectively.
  • Demonstrates antiproliferative effects in cell lines with high HER-2 levels.
  • Arrests cell cycle at G1-S phase in BT474 cells.
  • Inhibits MAPK and Akt phosphorylation, and induces p27.
  • Exhibits anticancer activities against HER-2 or EGFR expressing cancer cells in vivo.
  • Reduces tumor growth in xenograft models.