Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neratinib (maleate) | 915942-22-2 | MFCD30607264 | 99.7% | 673.11 | C34H33ClN6O7 | 10 MG
SDP

Catalog No. 5000297768
Encompass_Preferred

Neratinib maleate is the maleate salt of neratinib, an orally available, irreversible inhibitor selective for HER2 and EGFR kinases. Supplied for research use, it is used to probe HER2/EGFR signaling and to evaluate kinase inhibition in preclinical studies.

  • Irreversible HER2 and EGFR inhibitor with IC50s of 59 nM (HER2) and 92 nM (EGFR).
  • High purity (99.7%) suitable for analytical and biological assays.
  • Molecular weight 673.11 and formula C34H33ClN6O7.
  • Stable as a solid at 4°C; in solution: -80°C (6 months) or -20°C (1 month), protect from light.
  • Available in small milligram pack sizes for research applications.

Catalog No. 50-002-97768 Supplier Medchemexpress LLC Supplier No. HY32721B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neratinib maleate is the maleate salt of neratinib, an orally available, irreversible inhibitor selective for HER2 and EGFR kinases. Supplied for research use, it is used to probe HER2/EGFR signaling and to evaluate kinase inhibition in preclinical studies.

  • Irreversible HER2 and EGFR inhibitor with IC50s of 59 nM (HER2) and 92 nM (EGFR).
  • High purity (99.7%) suitable for analytical and biological assays.
  • Molecular weight 673.11 and formula C34H33ClN6O7.
  • Stable as a solid at 4°C; in solution: -80°C (6 months) or -20°C (1 month), protect from light.
  • Available in small milligram pack sizes for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.