Learn More
Medchemexpress LLC Neratinib (Standard) | 698387-09-6 | MFCD28073539 | 99.25% | 557.04 | 50 MG
SDP
Catalog No. 5000401468
Encompass_Preferred
Neratinib (Standard) is the analytical standard of Neratinib, intended for research and analytical applications. It is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as a reference standard.
  • Analytical standard for Neratinib.
  • Intended for research and analytical applications.
  • Orally available.
  • Irreversible.
  • Highly selective HER2 and EGFR inhibitor.
  • IC50s of 59 nM for HER2 and 92 nM for EGFR.
  • Used as a reference standard in HPLC, GC, and MS research experiments.
Catalog No. 50-004-01468 Supplier Medchemexpress LLC Supplier No. HY32721R50MG
Neratinib (Standard) is the analytical standard of Neratinib, intended for research and analytical applications. It is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively. This compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS as a reference standard.
  • Analytical standard for Neratinib.
  • Intended for research and analytical applications.
  • Orally available.
  • Irreversible.
  • Highly selective HER2 and EGFR inhibitor.
  • IC50s of 59 nM for HER2 and 92 nM for EGFR.
  • Used as a reference standard in HPLC, GC, and MS research experiments.