Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nerigliatin | 1245603-92-2 | 99.9% | 432.43 g/mol | C22H20N6O4 | 10 MG
SDP

Catalog No. 5000242219
Encompass_Preferred

Nerigliatin is a small-molecule glucokinase activator used for laboratory research into glucose-lowering mechanisms. It has demonstrated glucose-lowering efficacy in preclinical studies but has been associated with peripheral nerve degeneration in reported results. Intended for research use only and not for human or clinical applications.

  • Glucokinase activator for glucose-lowering research.
  • High purity (99.94%).
  • Molecular weight 432.43 g/mol.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Available in multiple small-scale solid quantities suitable for lab studies.
  • Not intended for clinical or human use; handle with appropriate safety precautions.

Catalog No. 50-002-42219 Supplier Medchemexpress LLC Supplier No. HY10832810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nerigliatin is a small-molecule glucokinase activator used for laboratory research into glucose-lowering mechanisms. It has demonstrated glucose-lowering efficacy in preclinical studies but has been associated with peripheral nerve degeneration in reported results. Intended for research use only and not for human or clinical applications.

  • Glucokinase activator for glucose-lowering research.
  • High purity (99.94%).
  • Molecular weight 432.43 g/mol.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Available in multiple small-scale solid quantities suitable for lab studies.
  • Not intended for clinical or human use; handle with appropriate safety precautions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.