Learn More
Medchemexpress LLC Nerigliatin | 1245603-92-2 | MFCD26960571 | 99.9% | 432.43 g·mol⁻¹ | C22H20N6O4 | 100 MG
SDP
Catalog No. 5000241838
Encompass_Preferred
Nerigliatin is a small-molecule glucokinase activator provided for laboratory research use. It is used in preclinical studies to evaluate glucose-lowering effects and glucokinase modulation. Supplied as a powdered compound, it is intended for in vitro and in vivo research applications; not for human or veterinary use.
  • Glucokinase activator for studying glucose metabolism.
  • High purity (≈99.9%) suitable for analytical and biological assays.
  • Chemical formula C22H20N6O4; molecular weight 432.43 g·mol⁻¹.
  • Available in multiple pack sizes including 100 MG.
  • For research use only; not intended for clinical or veterinary applications.
  • Follow safety data sheet for handling and storage recommendations.
Catalog No. 50-002-41838 Supplier Medchemexpress LLC Supplier No. HY108328100MG
Nerigliatin is a small-molecule glucokinase activator provided for laboratory research use. It is used in preclinical studies to evaluate glucose-lowering effects and glucokinase modulation. Supplied as a powdered compound, it is intended for in vitro and in vivo research applications; not for human or veterinary use.
  • Glucokinase activator for studying glucose metabolism.
  • High purity (≈99.9%) suitable for analytical and biological assays.
  • Chemical formula C22H20N6O4; molecular weight 432.43 g·mol⁻¹.
  • Available in multiple pack sizes including 100 MG.
  • For research use only; not intended for clinical or veterinary applications.
  • Follow safety data sheet for handling and storage recommendations.