Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neriifolin | 466-07-9 | 1 MG
SDP

Catalog No. 5000417383
Encompass_Preferred

Neriifolin is a CNS-penetrating cardiac glycoside that functions as an inhibitor of Na+, K+-ATPase. It targets beclin 1, inhibiting the formation of LC3-associated phagosomes and ameliorating the development of experimental autoimmune encephalomyelitis (EAE). This compound induces cell cycle arrest and apoptosis in human hepatocellular carcinoma HepG2 cells, making it suitable for laboratory research applications.

  • CNS-penetrating cardiac glycoside
  • Inhibitor of Na+, K+-ATPase
  • Targets beclin 1
  • Inhibits LC3-associated phagosome formation
  • Ameliorates experimental autoimmune encephalomyelitis (EAE) development
  • Induces cell cycle arrest and apoptosis in HepG2 cells
  • Purity: 99.70%
  • Appearance: Solid
  • Color: White to yellow
  • Molecular weight: 534.68
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-17383 Supplier Medchemexpress LLC Supplier No. HYN84411MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neriifolin is a CNS-penetrating cardiac glycoside that functions as an inhibitor of Na+, K+-ATPase. It targets beclin 1, inhibiting the formation of LC3-associated phagosomes and ameliorating the development of experimental autoimmune encephalomyelitis (EAE). This compound induces cell cycle arrest and apoptosis in human hepatocellular carcinoma HepG2 cells, making it suitable for laboratory research applications.

  • CNS-penetrating cardiac glycoside
  • Inhibitor of Na+, K+-ATPase
  • Targets beclin 1
  • Inhibits LC3-associated phagosome formation
  • Ameliorates experimental autoimmune encephalomyelitis (EAE) development
  • Induces cell cycle arrest and apoptosis in HepG2 cells
  • Purity: 99.70%
  • Appearance: Solid
  • Color: White to yellow
  • Molecular weight: 534.68
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.