Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NERL™ Reagent Grade Water

Catalog No. 23249580 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-249-580 32 oz.
23-249-579 1 gal.
23-249-581 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-249-580
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Improve control in critical diagnostic procedures by using this reagent grade water in place of deionized or distilled water.

Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ Reagent Grade Water in place of deionized or distilled water. This water meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.

  • Ideal for reconstituting chemistry or coagulation products, preparing analytical standards or rinsing delicate electrodes
  • Prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron
  • Essentially free from organic and inorganic particulate and soluble contaminants
  • Contains no preservatives
  • Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water
  • Certificate of Analysis is available online
TRUSTED_SUSTAINABILITY

Specifications

pH 5 to 7
Purity Grade Notes Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water. Certificate of Analysis is available online.
Grade USP/NF, CLRW
Quantity 32 oz.

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.