Learn More
Medchemexpress LLC Nerolidol | 7212-44-4 | 222.37 | 100 G
SDP
Catalog No. 5000415432
Encompass_Preferred
Nerolidol possesses multiple natural membrane activities, exhibiting anti-cancer, anti-inflammatory, antibacterial, and anti-insect properties. It suppresses parasitic activity and protects cells from damage to lipid and protein properties, as well as DNA.
  • Possesses anti-cancer activity
  • Possesses anti-inflammatory activity
  • Possesses antibacterial activity
  • Possesses anti-insect activity
  • Suppresses parasitic activity
  • Protects cells from damage to lipid and protein properties
  • Protects cells from DNA damage
  • Exhibits potent antioxidant properties
  • Shows neuroprotective effects
Catalog No. 50-004-15432 Supplier Medchemexpress LLC Supplier No. HYN1944100G
Nerolidol possesses multiple natural membrane activities, exhibiting anti-cancer, anti-inflammatory, antibacterial, and anti-insect properties. It suppresses parasitic activity and protects cells from damage to lipid and protein properties, as well as DNA.
  • Possesses anti-cancer activity
  • Possesses anti-inflammatory activity
  • Possesses antibacterial activity
  • Possesses anti-insect activity
  • Suppresses parasitic activity
  • Protects cells from damage to lipid and protein properties
  • Protects cells from DNA damage
  • Exhibits potent antioxidant properties
  • Shows neuroprotective effects