Learn More
Medchemexpress LLC Nerolidol | 7212-44-4 | MFCD00008911 | 222.37 | 25 G
SDP
Catalog No. 5000389517
Encompass_Preferred
Nerolidol exhibits multiple natural membrane activities, including anti-cancer, anti-inflammatory, antibacterial, and anti-insect properties. It helps protect cells from damage to lipids, proteins, and DNA by counteracting oxidative stress. This compound also shows potent antimicrobial activity against various strains of *Staphylococcus aureus*, including methicillin-resistant ones, and demonstrates anti-biofilm activity.
  • Possesses anti-cancer, anti-inflammatory, antibacterial, and anti-insect activity
  • Suppresses parasitic, bloodsucking, and bloodworm diseases
  • Protects cells from lipid, protein, and DNA damage
  • Exhibits potent antioxidant properties
  • Shows potent antimicrobial activity
  • Demonstrates anti-biofilm activity
  • Strong anti-fungal effect
  • Provides neuroprotective effects against oxidative stress
Catalog No. 50-003-89517 Supplier Medchemexpress LLC Supplier No. 25GHYN1944
Nerolidol exhibits multiple natural membrane activities, including anti-cancer, anti-inflammatory, antibacterial, and anti-insect properties. It helps protect cells from damage to lipids, proteins, and DNA by counteracting oxidative stress. This compound also shows potent antimicrobial activity against various strains of *Staphylococcus aureus*, including methicillin-resistant ones, and demonstrates anti-biofilm activity.
  • Possesses anti-cancer, anti-inflammatory, antibacterial, and anti-insect activity
  • Suppresses parasitic, bloodsucking, and bloodworm diseases
  • Protects cells from lipid, protein, and DNA damage
  • Exhibits potent antioxidant properties
  • Shows potent antimicrobial activity
  • Demonstrates anti-biofilm activity
  • Strong anti-fungal effect
  • Provides neuroprotective effects against oxidative stress