Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ Nerve Biopsy Kit with Glutaraldehyde 3% in 0.1 Phosphate Buffer
SDP

Catalog No. 21520239
Change view
Click to view available options
Quantity:
100 Kits/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-239 100 Kits/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-239 Supplier Cenmed™ Supplier No. NER002
Add to Cart
Add to Cart

Ensure precise specimen preservation during nerve biopsies using this Glutaraldehyde 3% in 0.1 Phosphate Buffer Kit.

  • Glutaraldehyde 3% in 0.1 Phosphate Buffer contained within a 5.2 mL vial, designed for optimal tissue fixation
  • Safely handle and transport specimens with our clear-orange biohazard specimen bags, measuring 6 x 9 in. and equipped with indicators for added safety assurance
  • 3 oz. Formalin, buffered to 10%, comes in a convenient half-filled container, providing reliable preservation for various laboratory applications
  • Specialized solutions maintain the integrity of your specimens, ensuring accurate results in research and experiments

Specifications

Product Type Specimen Collection Kit
Format Tube
Media Type Molecular Transport Media
Sterility Sterile
For Use With (Application) Collection, Transport, and Preservation of Biological Specimens
Quantity 100 Kits/Pk.
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.