Learn More
Enzo Life Sciences Nerve growth factor 7S (mouse) (100µg)
SDP
Catalog No. 502009354
Encompass_Preferred
Alternative name: NGF 7S. Purity: ≥98% (HPLC). Formulation: Lyophilized. Source: Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984). Long Term Storage: -20°C.
Catalog No. 50-200-9354 Supplier Enzo Life Sciences Supplier No. ALX521008C100
Alternative name: NGF 7S. Purity: ≥98% (HPLC). Formulation: Lyophilized. Source: Isolated from mouse submaxillary glands by a modification of the procedure described by T. Darling, et al.; Cell Culture Meth. Mol. Cell Biol. 4, 79 (1984). Long Term Storage: -20°C.