Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nerve growth factor beta (recombinant human) | 86923-98-0 | >95.0% | 100 UG
SDP

Catalog No. 5000290445
Encompass_Preferred

Recombinant human Beta-NGF (nerve growth factor-β) produced in HEK293 cells and supplied lyophilized. It supports neuronal survival and differentiation and is suitable for cell proliferation and signaling assays.

  • Expression system HEK293 cells.
  • Purity >95.0% by reducing SDS-PAGE.
  • Sequence residues S122-R239.
  • Formulation lyophilized from buffered solution (20 mM PB, 250 mM NaCl, pH 7.0 or 20 mM NaAC, 150 mM NaCl, pH 5.5).
  • Storage store at -20°C; stable two years from receipt; after reconstitution stable at 4°C for 1 week.
  • Bioactivity ED50 ≤2 ng/mL in TF-1 cell proliferation assay.

Catalog No. 50-002-90445 Supplier Medchemexpress LLC Supplier No. HYP7660100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Beta-NGF (nerve growth factor-β) produced in HEK293 cells and supplied lyophilized. It supports neuronal survival and differentiation and is suitable for cell proliferation and signaling assays.

  • Expression system HEK293 cells.
  • Purity >95.0% by reducing SDS-PAGE.
  • Sequence residues S122-R239.
  • Formulation lyophilized from buffered solution (20 mM PB, 250 mM NaCl, pH 7.0 or 20 mM NaAC, 150 mM NaCl, pH 5.5).
  • Storage store at -20°C; stable two years from receipt; after reconstitution stable at 4°C for 1 week.
  • Bioactivity ED50 ≤2 ng/mL in TF-1 cell proliferation assay.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.