Learn More
Cusabio Technology LLC Nerve growth factor NGF
Catalog No. 5000050542
Encompass_Preferred
This kit detects NGF (Nerve Growth Factor) in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method With a detection range of 0 78 pg/mL to 50 pg/mL and a sensitivity of 0 195 pg/mL it provides precise measurement of NGF levels in small sample volumes (50-100 L) The assay utilizes a specific capture and detection antibody pair generating results within 1-5 hours through colorimetric analysis at 450 nm Designed for research applications it ensures reliable performance in studying neurotrophic factors across biological matrices while maintaining consistency across replicates
Catalog No. 50-000-50542 Supplier Cusabio Technology LLC Supplier No. CSBE04685R
This kit detects NGF (Nerve Growth Factor) in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method With a detection range of 0 78 pg/mL to 50 pg/mL and a sensitivity of 0 195 pg/mL it provides precise measurement of NGF levels in small sample volumes (50-100 L) The assay utilizes a specific capture and detection antibody pair generating results within 1-5 hours through colorimetric analysis at 450 nm Designed for research applications it ensures reliable performance in studying neurotrophic factors across biological matrices while maintaining consistency across replicates