Learn More
Medchemexpress LLC HY-14775 5mg Medchemexpress, Nesbuvir CAS:691852-58-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY147755MG
Encompass_Preferred
Medchemexpress, HY-14775 5mg Nesbuvir CAS:691852-58-1 Nesbuvir is a nonnucleoside inhibitor of the hepatitis C virus (HCV) nonstructural protein 5B (NS5B) polymerase. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1638