Learn More
TARGETMOL CHEMICALS INC NESBUVIR 5MG
SDP
Catalog No. 502578831
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nesbuvir is a selective hepatitis C virus (HCV) nonstructural protein 5B (NS5B) RNA-dependent polymerase inhibitor.Nesbuvir has an IC50 of 9 nM against hepatocellular carcinoma cells containing 1b HCV replicons. purity: 98%
Catalog No. 50-257-8831 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ00905MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nesbuvir is a selective hepatitis C virus (HCV) nonstructural protein 5B (NS5B) RNA-dependent polymerase inhibitor.Nesbuvir has an IC50 of 9 nM against hepatocellular carcinoma cells containing 1b HCV replicons. purity: 98%