Learn More
Pyramex™ Neshoba™ Safety Eyewear
Catalog No. 19155628 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor, Mirror
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Coating
19-155-628 Indoor/Outdoor, Mirror -
19-155-623 Clear -
19-155-624 Clear Anti-fog, Scratch-resistant
19-155-625 Gray -
19-155-626 Gray Scratch-resistant
19-155-627 Amber -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-155-628 Supplier Pyramex™ Supplier No. S9780S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ultra lightweight spectacle (16 grams) with frameless design and thin flexible temples
  • Incredibly lightweight spectacle – only 16 grams
  • Frameless design with flexible temples provides comfortable and secure fit
  • 9.5 base curved lens provides excellent protection
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Certifications/Compliance (Europe) EN-166
    Lens Tint Indoor/Outdoor, Mirror
    Weight (Metric) 16 g
    Lens Material Polycarbonate