Learn More
Medchemexpress LLC NESS 0327 | 489.82 | C24H23Cl3N4O | 5MG
SDP
Catalog No. 5000308315
Encompass_Preferred
NESS 0327 is a cannabinoid antagonist with high selectivity for the cannabinoid CB1 receptor NESS 0327 is more than 60 000-fold selective for the CB1 receptor[1]
Catalog No. 50-003-08315 Supplier Medchemexpress LLC Supplier No. HY1171395MG
NESS 0327 is a cannabinoid antagonist with high selectivity for the cannabinoid CB1 receptor NESS 0327 is more than 60 000-fold selective for the CB1 receptor[1]