Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nessler Reagent, Reagents

Catalog No. 01337159
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-159 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-159 Supplier Reagents Holdings Llc Supplier No. C2253500500B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nessler Reagent, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Nessler Reagent formulated according to APHA 4500 standards for laboratory use.

  • Physical State: Liquid
  • Color: Light yellow
  • Grade: APHA
  • Composition: 79.65% Water, 12.49% Sodium Hydroxide, 4.37% Mercuric Iodide, 3.49% Potassium Iodide
  • Density: 1.27
  • Boiling Point: Approximately 102°C
  • Solubility: Miscible
  • Packaging: Amber Poly Container
  • Shelf Life: 2 Years
  • Storage: Ambient

Specifications

Color Light Yellow
Volume (Metric) 500 mL
Packaging Opaque HDPE
Quantity 500 mL
Size 500 ml
Sterility Non-sterile
Type Nessler Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.