Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nessler Reagent, StandARd, Macron Fine Chemicals™

Catalog No. MKH261100 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-H261-100 100 mL
MK-H261-4 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-H261-100 Supplier Avantor Supplier No. H26101
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7783-33-7
Molecular Formula H4HgI4K2
Molecular Weight (g/mol) 790.44
MDL Number MFCD00049656
InChI Key ZCPVFAXYZYZMBC-UHFFFAOYSA-N
Synonym nessler's reagent, mayer's reagent, potassium mercuric iodide, channing's solution, potassium tetraiodomercurate ii, nessler reagent, caswell no. 695, potassium iodomercurate, mercury potassium iodide, mercuric potassium iodide
PubChem CID 24542
ChEBI CHEBI:51568
IUPAC Name dipotassium tetraiodomercurydiuide
SMILES [K+].[K+].I.I.I.I.[Hg--]

Specifications

Quantity 100 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.