Learn More
Hach Company Nessler Reagent, 100 mL MDB
Supplier:  Hach Company 2119432
Encompass
For ammonia nitrogen determination by the Nessler method (Hach method 8038). EPA accepted for wastewater analysis (distillation required). Range: 0.02-2.50 mg/L NH3-N. Also used in visual test kits - alternate ranges apply. 100 mL bottle.
Catalog No. 50-199-5445