Learn More
Hach Company Nessler Reagent, 500 mL
Supplier:  Hach Company 2119449
Encompass
For ammonia nitrogen determination by the Nessler method (Hach method 8038). EPA accepted for wastewater analysis (distillation required). Range: 0.02-2.50 mg/L NH3-N. 500 mL bottle. *Item cannot be shipped to Louisianna due to a state restriction
Catalog No. 50-199-5446