Learn More
Nesslers Reagent, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| Chemical Name or Material | Nesslers Reagent |
| Color | Yellow |
| Name Note | For Nitrogen (Ammonia), Nitrogen (Organic) |
| CAS Max % | 67, 16, 10, 7 |
| CAS | 1310-73-2, 7774-29-0, 7681-11-0 |
| Linear Formula | HgI2 |
| UN Number | UN2922 |
| Solubility Information | Soluble in water |
| Formula Weight | 454.4 |
| Grade | Certified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.