Learn More
NEST SCIENTIFIC INC Nest BioFactory 5 Chamber; 2 Wide Plug Seal Caps; Total culture area: 3175cm2; TC; sterile, 1/pk, 4/cs (with 8 sterile wide vent caps included separately)
Catalog No. NC2887686
Encompass_Preferred
Nest BioFactory™ Systems are compact, multi-layer, single-use cell culture systems designed for easy scale-up cell culture applications, such as production of vaccines, monoclonal antibodies or pharmaceuticals. Versatile, easy-to-use systems for small scale research or commercial production with reduced contamination risk. With ISO9001&13485 certified, 5 layers BioFactory™ are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials. • Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.• Individually packaged in double-layer sterile bags.• TC treated• Clear lot number for batch traceability.• Ultrasonic welding without extrinsic ingredients.• Vent caps with 0.22 μm hydrophobic filters for gas exchange without contamination.• Wide mouth is applicable to pour culture medium directly. Narrow mouth is applicable to operate with the aseptic pipeline.• Applicable to automatic machine.
Catalog No. NC2887686 Supplier NEST SCIENTIFIC INC Supplier No. 771204
Nest BioFactory™ Systems are compact, multi-layer, single-use cell culture systems designed for easy scale-up cell culture applications, such as production of vaccines, monoclonal antibodies or pharmaceuticals. Versatile, easy-to-use systems for small scale research or commercial production with reduced contamination risk. With ISO9001&13485 certified, 5 layers BioFactory™ are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials. • Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.• Individually packaged in double-layer sterile bags.• TC treated• Clear lot number for batch traceability.• Ultrasonic welding without extrinsic ingredients.• Vent caps with 0.22 μm hydrophobic filters for gas exchange without contamination.• Wide mouth is applicable to pour culture medium directly. Narrow mouth is applicable to operate with the aseptic pipeline.• Applicable to automatic machine.