Learn More
Bellco Glass, Inc. Nest Cell Culture Chamber Slides 8 well w/glass slide White 0.98 cm2 0.20.6ml 6/pk 12/cs
SDP
Catalog No. 502329825
Encompass_Preferred
Nest Cell Culture Chamber Slides 8 well w/glass slide White 0.98 cm2 0.20.6ml 6/pk 12/cs
Catalog No. 50-232-9825 Supplier Bellco Glass, Inc. Supplier No. 451400126
Nest Cell Culture Chamber Slides 8 well w/glass slide White 0.98 cm2 0.20.6ml 6/pk 12/cs