Learn More
Lakeside Manufacturing Inc Nesting Instrument Table, Stainless Steel, 20" x 40", 200-Lb. Capacity
Catalog No. 502884464
Encompass_Preferred
Nesting Instrument Tables by Lakeside Manufacturing save space in crowded operating rooms and provide surgeons streamlined access to supplies. The Lakeside 8383 Nesting Instrument Table measuring 20" x 40" has a weight capacity of 200-Lb. This Lakeside instrument table's stainless steel top is easy to clean and sanitize. Its 1” (24) square 16-gauge stainless steel legs offer superior strength during transport. This table's all welded construction increases durability and makes surface smoother for cleaning. Its 3” (76) locking casters ensure table is stationary when needed. Sits at a comfortable working height of 38" allows easy for access to medical supplies and equipment. Its nesting tables keep workspaces clutter free, reducing accidents. 200-Lb. (91kg) weight capacity holds light duty medical supplies and equipment. Reinforcements supply extra security for larger models. Endless variations and feature combinations. Superior lead times. Made in the USA.
Catalog No. 50-288-4464 Supplier Lakeside Manufacturing Inc Supplier No. 8383
Nesting Instrument Tables by Lakeside Manufacturing save space in crowded operating rooms and provide surgeons streamlined access to supplies. The Lakeside 8383 Nesting Instrument Table measuring 20" x 40" has a weight capacity of 200-Lb. This Lakeside instrument table's stainless steel top is easy to clean and sanitize. Its 1” (24) square 16-gauge stainless steel legs offer superior strength during transport. This table's all welded construction increases durability and makes surface smoother for cleaning. Its 3” (76) locking casters ensure table is stationary when needed. Sits at a comfortable working height of 38" allows easy for access to medical supplies and equipment. Its nesting tables keep workspaces clutter free, reducing accidents. 200-Lb. (91kg) weight capacity holds light duty medical supplies and equipment. Reinforcements supply extra security for larger models. Endless variations and feature combinations. Superior lead times. Made in the USA.