Learn More
Electron Microscopy Sciences Nesting Sample Cups, Capacity: 2 mL. Dimensions: 16mm x 30mm., 1000/EA
SDP
Catalog No. 502090613
Encompass_Preferred
These nesting sample cups are ideal for when a smaller sampling is nesessary. Use them together with other test tubes or original blood collection tubes. Simply transfer the sample from the original collection tube into the nesting cup. Then place the nesting cup back into the original collection tube. Using the procedure allows you to make use of the original tube's label and/or barcode. Ideal for use on direct sampling machines as Dade Dimension, Hitachi 747 and more. Capacity: 2 mL. Dimensions: 16mm x 30mm .
Catalog No. 50-209-0613 Supplier Electron Microscopy Sciences Supplier No. 6099416
These nesting sample cups are ideal for when a smaller sampling is nesessary. Use them together with other test tubes or original blood collection tubes. Simply transfer the sample from the original collection tube into the nesting cup. Then place the nesting cup back into the original collection tube. Using the procedure allows you to make use of the original tube's label and/or barcode. Ideal for use on direct sampling machines as Dade Dimension, Hitachi 747 and more. Capacity: 2 mL. Dimensions: 16mm x 30mm .