Learn More
Medchemexpress LLC Nesuparib | 2055357-64-5 | 99.5% | 400.48 g·mol⁻1 | C23H24N6O | 5 MG
SDP
Catalog No. 5000221732
Encompass_Preferred
Nesuparib is an orally bioavailable small-molecule inhibitor of PARP1/2 and tankyrase1/2, provided as a high-purity solid for preclinical and in vitro research use.
  • Orally bioavailable small molecule.
  • Inhibits PARP and tankyrase enzymes.
  • High purity (~99.5%) solid suitable for assays.
  • Molecular formula C23H24N6O; molecular weight 400.48 g·mol⁻1.
  • Supplied as a 5 mg solid pack for laboratory research.
  • Intended for research use only, not for human therapeutic use.
Catalog No. 50-002-21732 Supplier Medchemexpress LLC Supplier No. HY1455845MG
Nesuparib is an orally bioavailable small-molecule inhibitor of PARP1/2 and tankyrase1/2, provided as a high-purity solid for preclinical and in vitro research use.
  • Orally bioavailable small molecule.
  • Inhibits PARP and tankyrase enzymes.
  • High purity (~99.5%) solid suitable for assays.
  • Molecular formula C23H24N6O; molecular weight 400.48 g·mol⁻1.
  • Supplied as a 5 mg solid pack for laboratory research.
  • Intended for research use only, not for human therapeutic use.