Learn More
Medchemexpress LLC Nesuparib | 2055357-64-5 | 99.5% | C23H24N6O | 10MG
SDP
Catalog No. 5000258725
Encompass_Preferred
Nesuparib is an orally active small-molecule inhibitor of PARP1/2 and tankyrase 1/2 supplied for laboratory research. It exhibits potent inhibition of its targets and is provided as a high-purity compound for use in biochemical and pharmacology studies. Not for clinical or human use.
Catalog No. 50-002-58725 Supplier Medchemexpress LLC Supplier No. HY14558410MG
Nesuparib is an orally active small-molecule inhibitor of PARP1/2 and tankyrase 1/2 supplied for laboratory research. It exhibits potent inhibition of its targets and is provided as a high-purity compound for use in biochemical and pharmacology studies. Not for clinical or human use.