Learn More
Medchemexpress LLC HY-124957 10mg Medchemexpress, β-NETA CAS:31059-54-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12495710MG
Encompass_Preferred
Medchemexpress, HY-124957 10mg β-NETA CAS:31059-54-8 β-NETA is a potent and noncompetitive choline acetyltransferase (ChA; IC50=76 μM) and cholinesterase (ChE; IC50=40 µM) inhibitor. β-NETA weakly inhibits acetylcholinesterase (AChE; IC50=1 mM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1061