Learn More
Medchemexpress LLC Netarsudil hydrochloride | 1253952-02-1 | 99.9% | 100 MG
SDP
Catalog No. 5000339276
Encompass_Preferred
Netarsudil hydrochloride is a competitive inhibitor of Rho-associated protein kinases (ROCK I and ROCK II) and a reversible inhibitor of the norepinephrine transporter (NET). It is primarily used in the study of ocular hypertension and primary open-angle glaucoma, reducing intraocular pressure by inhibiting ROCK and reducing aqueous humor production by inhibiting NET.
  • Inhibits ROCK I and ROCK II
  • Reversible inhibitor of norepinephrine transporter (NET)
  • Reduces intraocular pressure
  • Increases aqueous humor outflow ease
  • Reduces aqueous humor production
  • Causes relaxation of trabecular meshwork cells
  • Dilation of episcleral veins
  • Metabolizes to active compound Netarsudil-M1
Catalog No. 50-003-39276 Supplier Medchemexpress LLC Supplier No. HY12798B100MG
Netarsudil hydrochloride is a competitive inhibitor of Rho-associated protein kinases (ROCK I and ROCK II) and a reversible inhibitor of the norepinephrine transporter (NET). It is primarily used in the study of ocular hypertension and primary open-angle glaucoma, reducing intraocular pressure by inhibiting ROCK and reducing aqueous humor production by inhibiting NET.
  • Inhibits ROCK I and ROCK II
  • Reversible inhibitor of norepinephrine transporter (NET)
  • Reduces intraocular pressure
  • Increases aqueous humor outflow ease
  • Reduces aqueous humor production
  • Causes relaxation of trabecular meshwork cells
  • Dilation of episcleral veins
  • Metabolizes to active compound Netarsudil-M1