Learn More
Cayman Chemical N-ethyl Perfluorooctanesulfonamide, Purity: ≥85%, CAS: 4151-50-2, 50 mg
SDP
Catalog No. 502915413
Encompass_Preferred
N-ethyl Perfluorooctanesulfonamide (N-ethyl FOSA) is a perfluoroalkyl substance (PFAS) and an insecticide.{49815,49816} It decreases the coupling efficiency, increases the rate of state four respiration, and uncouples mitochondrial oxidative phosphorylation in rat liver mitochondria when used at a concentration of 6.25 µM.{49817} N-ethyl FOSA reduces foraging activity of leaf-cutting ants (A. lundi) and reduces colony survival when applied in colonies as a 0.3% bait.{49816} It has been found in human blood and Arctic air samples.{49815,49818}
Catalog No. 50-291-5413 Supplier Cayman Chemical Supplier No. 3726350 MG
N-ethyl Perfluorooctanesulfonamide (N-ethyl FOSA) is a perfluoroalkyl substance (PFAS) and an insecticide.{49815,49816} It decreases the coupling efficiency, increases the rate of state four respiration, and uncouples mitochondrial oxidative phosphorylation in rat liver mitochondria when used at a concentration of 6.25 µM.{49817} N-ethyl FOSA reduces foraging activity of leaf-cutting ants (A. lundi) and reduces colony survival when applied in colonies as a 0.3% bait.{49816} It has been found in human blood and Arctic air samples.{49815,49818}