Learn More
Cayman Chemical Nethyl PRfluoroctnsulfon 100mg
SDP
Supplier:  Cayman Chemical 37916100 MG
Encompass_Preferred
A perfluoroalkyl substance; decreases maternal body weight and food intake in pregnant rats and induces abortions and decreases fetal weight in pregnant rabbits at 10 mg/kg; has been found in landfill leachate and the atmosphere
Catalog No. 50-291-7457