Learn More
Medchemexpress LLC Neticonazole hydrochloride | 130773-02-3 | 99.96% | 25 MG
SDP
Catalog No. 5000341411
Encompass_Preferred
Neticonazole hydrochloride is an imidazole derivative that acts as a potent and long-acting antifungal agent. It also demonstrates anti-infection and anti-cancer effects. This product has been shown to inhibit exosome release and decrease levels of Alix, Rab27a, and nSMase2 in C4-2B cells. It also inhibits p-ERK levels. In vivo research indicates it can significantly improve survival in mice with colorectal cancer xenograft tumors.
  • Potent and long-acting antifungal agent
  • Demonstrates anti-infection and anti-cancer effects
  • Inhibits exosome release
  • Decreases Alix, Rab27a, and nSMase2 levels
  • Inhibits p-ERK levels
  • Acts as an orally active exosome biogenesis and secretion inhibitor
  • Improves survival in intestinal dysbacteriosis mice with colorectal cancer xenograft tumors
Catalog No. 50-003-41411 Supplier Medchemexpress LLC Supplier No. 25MGHY128365
Neticonazole hydrochloride is an imidazole derivative that acts as a potent and long-acting antifungal agent. It also demonstrates anti-infection and anti-cancer effects. This product has been shown to inhibit exosome release and decrease levels of Alix, Rab27a, and nSMase2 in C4-2B cells. It also inhibits p-ERK levels. In vivo research indicates it can significantly improve survival in mice with colorectal cancer xenograft tumors.
  • Potent and long-acting antifungal agent
  • Demonstrates anti-infection and anti-cancer effects
  • Inhibits exosome release
  • Decreases Alix, Rab27a, and nSMase2 levels
  • Inhibits p-ERK levels
  • Acts as an orally active exosome biogenesis and secretion inhibitor
  • Improves survival in intestinal dysbacteriosis mice with colorectal cancer xenograft tumors