Learn More
TARGETMOL CHEMICALS INC Neticonazole Hydrochloride
SDP
Catalog No. 502878897
Encompass_Preferred
Also available in 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Neticonazole Hydrochloride is an imidazole antifungal for the treatment of fungal skin infections. Neticonazole is reported as a potent exosome inhibitor which can block the activity of exosomes. Purity 99.84%
Catalog No. 50-287-8897 Supplier TARGETMOL CHEMICALS INC Supplier No. T4985100MG
Also available in 25 mg, 50 mg, 200 mg and bulk. Please contact Fisher for quotes. Neticonazole Hydrochloride is an imidazole antifungal for the treatment of fungal skin infections. Neticonazole is reported as a potent exosome inhibitor which can block the activity of exosomes. Purity 99.84%