Learn More
Medchemexpress LLC HY-A0086 10mg Medchemexpress, Netilmicin (sulfate) CAS:56391-57-2 Purity:>98%
SDP
Catalog No. 502082321
Encompass_Preferred
Medchemexpress, HY-A0086 10mg Netilmicin (sulfate) CAS:56391-57-2 Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2321 Supplier Medchemexpress LLC Supplier No. HYA008610MG
Medchemexpress, HY-A0086 10mg Netilmicin (sulfate) CAS:56391-57-2 Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.