Learn More
Apexbio Technology LLC Netilmicin Sulfate 56391-57-2 5mg
SDP
Catalog No. 5000094141
Encompass_Preferred
Netilmicin Sulfate is a small-molecule antibiotic targeting the bacterial 30S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby disrupting bacterial growth Netilmicin Sulfate exerts its biological activity primarily through binding to the bacterial 30S ribosomal subunit resulting in inhibition of protein synthesis Based on these pharmacological properties Netilmicin Sulfate holds research potential in microbiological studies of antimicrobial resistance profiles bacterial infection mechanisms resistance development assays and translational research focused on bacterial ribosomal function
Catalog No. 50-000-94141 Supplier Apexbio Technology LLC Supplier No. B17965
Netilmicin Sulfate is a small-molecule antibiotic targeting the bacterial 30S ribosomal subunit It is designed to inhibit bacterial protein synthesis thereby disrupting bacterial growth Netilmicin Sulfate exerts its biological activity primarily through binding to the bacterial 30S ribosomal subunit resulting in inhibition of protein synthesis Based on these pharmacological properties Netilmicin Sulfate holds research potential in microbiological studies of antimicrobial resistance profiles bacterial infection mechanisms resistance development assays and translational research focused on bacterial ribosomal function